• USA Home
  • Search Results

  • sodiumchlorate10644777509911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium chlorate

3 Products
Image of Sodium chlorate
CAS Number:
Linear Formula:
 NaClO3
Molecular Weight:
 106.44
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Solubility
  Potency
  Physical Form
Show More