• USA Home
  • Search Results

  • sodiumchloride5844764714511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium chloride

34 Products
Synonym:   Halite
Image of Sodium chloride
CAS Number:
Linear Formula:
 NaCl
Molecular Weight:
 58.44
 Beilstein/REAXYS Number:
 3534976
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Absorption
  UV-Absorption Values
  UV-Absorption Condition
  Package quantity
  Anal. Standard Form
  Ice Shipping
  Restriction
  Suitability
  Particle Dimension
  Manufacturer designation
  Loss
  Quality clarifier
  Purity Grade
  Product Line
  Bulk Density
  Application(s)
  Potency
  Agency/Method
  Physical Form
  Vapor pressure
  Solubility
  Assay
  pH Value
Show More

Product #
Description
S7653 Sodium chloride, BioXtra, ≥99.5% (AT),
S9888 Sodium chloride, ACS reagent, ≥99.0%,
S3014 Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration),
S5886 Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%,
S9625 Sodium chloride, ReagentPlus®, ≥99%,
1.06404 Sodium chloride, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
746398 Sodium chloride, anhydrous, Redi-Dri, free-flowing, ACS reagent, ≥99%,
S1679 Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%,
31434 Sodium chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%,
71376 Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT),
1.06400 Sodium chloride, EMPROVE® ESSENTIAL Ph Eur,BP,USP,
793566 Sodium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%,
1.16224 Sodium chloride, low in endotoxins suitable for use as excipient EMPROVE® exp Ph Eur,BP,JP,USP,
1.37017 Sodium chloride, EMPROVE® EXPERT Ph Eur,BP,JP,USP,
71380 Sodium chloride, puriss. p.a., ≥99.5% (AT),
204439 Sodium chloride, 99.999% trace metals basis,
13423 Sodium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al,
PHR1321 Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material,
71387 Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%,
S6191 Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture,
1.02406 Sodium chloride, volumetric standard, secondary reference material for argentometry, traceable to NIST SRM Certipur® Reag. Ph Eur,Reag. USP,
1.06406 Sodium chloride, 99.99 Suprapur®,
SX0420 Sodium chloride, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
71383 Sodium chloride, tested according to Ph Eur,
NIST919B Sodium chloride, NIST® SRM® 919b,
567440 Sodium chloride, ACS Reagent Grade.,
5.43832 Sodium chloride, for HPLC LiChropur,
71382 Sodium chloride, puriss. p.a., ≥99.5% (AT), powder or crystals,
450006 Sodium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis,
1613804 Sodium chloride, United States Pharmacopeia (USP) Reference Standard,
1.06444 Sodium chloride, EMPROVE® API Ph Eur,BP,JP,USP,
378860 Sodium chloride, random crystals, optical grade, 99.9% trace metals basis,
S6150 Sodium chloride, tablet,
RDD041 Sodium chloride, Redi-Dri, ≥99%, DNase, RNase, and protease (none detected), free-flowing,