• USA Home
  • Search Results

  • sodiumchloridesolution5844764714511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium chloride solution

7 Products
Image of Sodium chloride solution
CAS Number:
Linear Formula:
 NaCl
Molecular Weight:
 58.44
 Beilstein/REAXYS Number:
 3534976
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Restriction
  Suitability
  pH Value
  UV-Absorption Values
  Product Line
  ID clarifier
  Special Grade
  Sterilization
  Density
Show More