• USA Home
  • Search Results

  • sodiumcholatehydrate43055anhydrousbasis20698687011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium cholate hydrate

5 Products
Synonym:   3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt, Cholalic acid sodium salt
Image of Sodium cholate hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C24H39NaO5 · xH2O
Molecular Weight:
 430.55 (anhydrous basis)
 Beilstein/REAXYS Number:
 3582354
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More