• USA Home
  • Search Results

  • sodiumcitratedihydrate29410613204311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium citrate dihydrate

1 Products
Synonym:   Citric acid trisodium salt dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate dihydrate
Image of Sodium citrate dihydrate
CAS Number:
Linear Formula:
 HOC(COONa)(CH2COONa)2 · 2H2O
Molecular Weight:
 294.10
 Beilstein/REAXYS Number:
 6104939
 EC Number:
 FEMA Number:
 3026
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More