• USA Home
  • Search Results

  • sodiumcyanate650191761311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium cyanate

1 Products
Synonym:   Sodium isocyanate, Zassol
Image of Sodium cyanate
CAS Number:
Linear Formula:
 NaOCN
Molecular Weight:
 65.01
 Beilstein/REAXYS Number:
 3655041
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 38060512
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
185086 Sodium cyanate, 96%,