• USA Home
  • Search Results

  • sodiumcyanide490114333911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium cyanide

3 Products
Image of Sodium cyanide
CAS Number:
Empirical Formula (Hill Notation):
 CNNa
Molecular Weight:
 49.01
 EC Index Number
 205-599-4
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Purity Grade
  Solubility
  Special Grade
Show More