• USA Home
  • Search Results

  • sodiumcyanoborohydride62842589560711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium cyanoborohydride

3 Products
Synonym:   Sodium cyanotrihydridoborate
Image of Sodium cyanoborohydride
CAS Number:
Linear Formula:
 NaBH3CN
Molecular Weight:
 62.84
 Beilstein Registry Number:
 4152656
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Special Grade
  Assay
Show More