• USA Home
  • Search Results

  • sodiumcyclamate2012213905911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium cyclamate

4 Products
Image of Sodium cyclamate
CAS Number:
Empirical Formula (Hill Notation):
 C6H12NNaO3S
Molecular Weight:
 201.22
 EC Index Number
 205-348-9
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
Show More