• USA Home
  • Search Results

  • sodiumdeoxycholatemonohydrate4325714522492611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium deoxycholate monohydrate

2 Products
Synonym:   3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt
Image of Sodium deoxycholate monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C24H39NaO4 · H2O
Molecular Weight:
 432.57
 Beilstein/REAXYS Number:
 3643164
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  pH Value
  UV-Absorption Values
  Titertest
  Solubility
  Product Line
  Molecular weight
  ID clarifier
Show More