• USA Home
  • Search Results

  • sodiumdllactatesolution112067217311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium DL-lactate solution

3 Products
Synonym:   DL-Lactic acid sodium salt
Image of Sodium <SC>DL</SC>-lactate solution
CAS Number:
Linear Formula:
 CH3CH(OH)COONa
Molecular Weight:
 112.06
 Beilstein/REAXYS Number:
 4332999
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Application(s)
  Product Line
  Physical Form
  Biological Source
Show More