• USA Home
  • Search Results

  • sodiumdodecylbenzenesulfonate348482515530011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium dodecylbenzenesulfonate

2 Products
Synonym:   Dodecylbenzenesulfonic acid sodium salt, SDBS
Image of Sodium dodecylbenzenesulfonate
CAS Number:
Linear Formula:
 CH3(CH2)11C6H4SO3Na
Molecular Weight:
 348.48
 Beilstein/REAXYS Number:
 4171051
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More