• USA Home
  • Search Results

  • sodiumdodecylsulfatesolution2883815121311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium dodecyl sulfate solution

2 Products
Synonym:   Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate solution
Image of Sodium dodecyl sulfate solution
CAS Number:
Linear Formula:
 CH3(CH2)11OSO3Na
Molecular Weight:
 288.38
 Beilstein/REAXYS Number:
 3599286
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 39093306
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
  UV-Absorption Condition
  Product Line
  Molecular weight
  ID clarifier
  Density
  Concentration
  UV-Absorption Values
Show More