• USA Home
  • Search Results

  • sodiumformate13c18o273009515017711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium formate-13C,18O2

1 Products
Synonym:   Formic acid-13C,18O2 sodium salt
Image of Sodium formate-<SUP>13</SUP>C,<SUP>18</SUP>O<SUB>2</SUB>
CAS Number:
Linear Formula:
 H13C18O2Na
Molecular Weight:
 73.00
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
Show More