• USA Home
  • Search Results

  • sodiumhexafluorosilicate188061689385911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium hexafluorosilicate

1 Products
Image of Sodium hexafluorosilicate
CAS Number:
Linear Formula:
 Na2SiF6
Molecular Weight:
 188.06
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
250171 Sodium hexafluorosilicate,