• USA Home
  • Search Results

  • sodiumhexanitrocobaltateiii403941360098111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium hexanitrocobaltate(III)

3 Products
Synonym:   Cobalt(III) sodium nitrite, Sodium cobaltinitrite, Sodium hexanitrocobaltate(III)
Image of Sodium hexanitrocobaltate(III)
CAS Number:
Linear Formula:
 Na3[Co(NO2)6]
Molecular Weight:
 403.94
 EC Index Number
 237-077-7
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More