• USA Home
  • Search Results

  • sodiumhydrogentartrate1720752694311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium hydrogentartrate

1 Products
Synonym:   L(+)-Tartaric acid monosodium salt, Sodium bitartrate, Tartaric acid monosodium salt
Image of Sodium hydrogentartrate
CAS Number:
Linear Formula:
 NaO2CCH(OH)CH(OH)CO2H
Molecular Weight:
 172.07
 Beilstein/REAXYS Number:
 6099125
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More