• USA Home
  • Search Results

  • sodiumhydroxide4000131073211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium hydroxide

21 Products
Synonym:   ‘Caustic soda’
Image of Sodium hydroxide
CAS Number:
Linear Formula:
 NaOH
Molecular Weight:
 40.00
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  UV-Absorption Values
  Restriction
  Quality clarifier
  Purity Grade
  pH Value
  Agency/Method
  Product Line
  Application(s)
  Special Grade
  Density
  Solubility
  Physical Form
Show More

Product #
Description
S8045 Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous),
221465 Sodium hydroxide, ACS reagent, ≥97.0%, pellets,
S5881 Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous),
1.06498 Sodium hydroxide, pellets for analysis EMSURE®,
06203 Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets,
30620 Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets,
306576 Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis,
655104 Sodium hydroxide, reagent grade, 97%, powder,
795429 Sodium hydroxide, anhydrous, free-flowing, pellets, Redi-Dri, ACS reagent, ≥97%,
1.06469 Sodium hydroxide, pellets for analysis (max. 0.02% K) EMSURE® ACS,Reag. Ph Eur,ISO,
1.37020 Sodium hydroxide, pellets suitable for the biopharmaceutical production EMPROVE® bio Ph Eur,BP,JP,NF,
1.06462 Sodium hydroxide, pellets EMPLURA®,
71690 Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets,
71687 Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets,
367176 Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%,
484024 Sodium hydroxide, reagent grade, 97%, flakes,
1.60309 Sodium hydroxide, 0.01 M NaOH (0.01 N), Titripur®,
757527 Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis,
1.06495 Sodium hydroxide,
1.06467 Sodium hydroxide, granulated EMPLURA®,
901915 Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets,