• USA Home
  • Search Results

  • sodiumhydroxidepellets4000131073211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium hydroxide pellets

3 Products
Synonym:   ‘Caustic soda’, Soda caustic, Sodium hydroxide
Image of Sodium hydroxide pellets
CAS Number:
Linear Formula:
 NaOH
Molecular Weight:
 40.00
 E Number:
 E524
 EC Index Number:
 215-185-5
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Application(s)
  Ice Shipping
  Physical Form
  pH Value
  Solubility
  Density
  Assay
Show More