• USA Home
  • Search Results

  • sodiumhydroxidesolution4000131073211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium hydroxide solution

24 Products
Image of Sodium hydroxide solution
CAS Number:
Linear Formula:
 NaOH
Molecular Weight:
 40.00
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  UV-Absorption Condition
  Quality clarifier
  Product Line
  Composite Dimension
  Assay
  Special Grade
  Agency/Method
  Restriction
  UV-Absorption Values
  Reaction Suitability
  pH Value
  Application(s)
  Density
  Physical Form
Show More

Product #
Description
415413 Sodium hydroxide solution, 50% in H2O,
72068 Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O,
S2770 Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture,
1.09137 Sodium hydroxide solution, c(NaOH) = 1 mol/l (1 N) Titripur® Reag. Ph Eur,Reag. USP,
1.09141 Sodium hydroxide solution, c(NaOH) = 0.1 mol/l (0.1 N) Titripur® Reag. Ph Eur,
S8263 Sodium hydroxide solution, 5.0 M,
72064 Sodium hydroxide solution, 49-51% in water, eluent for IC,
1.09138 Sodium hydroxide solution, c(NaOH) = 0.5 mol/l (0.5 N) Titripur®,
79724 Sodium hydroxide solution, 1 M, for HPCE,
1.09136 Sodium hydroxide solution, c(NaOH) = 2 mol/l (2 N) Titripur®,
1.09959 Sodium hydroxide solution, for 1000 ml, c(NaOH) = 0.1 mol/l (0.1 N) Titrisol®,
1.09956 Sodium hydroxide solution, for 1000 ml, c(NaOH) = 1 mol/l (1 N) Titrisol®,
05211 Sodium hydroxide solution, purum, ≥32%,
1.09139 Sodium hydroxide solution, c(NaOH) = 0.25 mol/l (0.25 N) Titripur®,
1.09140 Sodium hydroxide solution, c(NaOH) = 0.2 mol/l (0.2 N) Titripur®,
1.09961 Sodium hydroxide solution, for 1000 ml, c(NaOH) = 0.01 mol/l (0.01 N) Titrisol®,
1.11584 Sodium hydroxide solution, c(NaOH) = 4 mol/l (4 N) Titripur®,
1.09142 Sodium hydroxide solution, c(NaOH) = 0.02 mol/l (0.02 N) Titripur®,
1.09957 Sodium hydroxide solution, for 1000 ml, c(NaOH) = 0.5 mol/l (0.5 N) Titrisol®,
69413 Sodium hydroxide solution, for HPLC, LiChropur, 49.0-51.0%,
150705 Sodium hydroxide solution, 0.1 M, Set incl. Absorber cartridge with soda lime, for 3S adapter technology,
150706 Sodium hydroxide solution, 1 M, Set incl. Absorber cartridge with soda lime, for 3S adapter technology,
1.09958 Sodium hydroxide solution, for 1000 ml, c(NaOH) = 0.25 mol/l (0.25 N) Titrisol®,
1.05595 Sodium hydroxide solution, c(NaOH) = 0.33 mol/l (1/3 N) Titripur®,