• USA Home
  • Search Results

  • sodiumhydroxidesolutionabout324000131073211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium hydroxide solution about 32%

2 Products
Synonym:   Sodium hydroxide solution
Image of Sodium hydroxide solution about 32%
CAS Number:
Linear Formula:
 NaOH
Molecular Weight:
 40.00
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Density
  pH Value
  Vapor pressure
  Melting Point
Show More