• USA Home
  • Search Results

  • sodiumhypophosphitehydrate8798anhydrousbasis12333367511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium hypophosphite hydrate

1 Products
Image of Sodium hypophosphite hydrate
CAS Number:
Linear Formula:
 NaH2PO2 · xH2O
Molecular Weight:
 87.98 (anhydrous basis)
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More