• USA Home
  • Search Results

  • sodiumiodide14989768182511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium iodide

13 Products
Image of Sodium iodide
CAS Number:
Linear Formula:
 NaI
Molecular Weight:
 149.89
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Purity Grade
  Loss
  Physical Form
  Quality clarifier
  Potency
  Density
  Bulk Density
  Agency/Method
  Vapor pressure
  Melting Point
  Solubility
  pH Value
  Assay
Show More

Product #
Description
383112 Sodium iodide, ACS reagent, ≥99.5%,
409286 Sodium iodide, 99.999% trace metals basis,
793558 Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%,
217638 Sodium iodide, ReagentPlus®, ≥99%,
1.06523 Sodium iodide, for analysis EMSURE® ACS,Reag. Ph Eur,
746371 Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%,
229911 Sodium iodide, ≥99.99% trace metals basis,
03129 Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance),
71710 Sodium iodide, puriss. p.a., ≥99.0% (AT),
1.06519 Sodium iodide, 99.99 Suprapur®,
1.06520 Sodium iodide, EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,
439681 Sodium iodide, anhydrous, beads, −10 mesh, 99.999% trace metals basis,
SX0625 Sodium Iodide, Meets ACS Specifications GR ACS,