• USA Home
  • Search Results

  • sodiumlaurylsulfate2883815121311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium lauryl sulfate

2 Products
Synonym:   Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium dodecyl sulfate, Sodium lauryl sulfate
Image of Sodium lauryl sulfate
CAS Number:
Linear Formula:
 CH3(CH2)11OSO3Na
Molecular Weight:
 288.38
 Beilstein/REAXYS Number:
 3599286
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More