• USA Home
  • Search Results

  • sodiummetaboratetetrahydrate137861055576711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium metaborate tetrahydrate

1 Products
Synonym:   Boric acid, sodium salt, tetrahydrate
Image of Sodium metaborate tetrahydrate
CAS Number:
Linear Formula:
 NaBO2 · 4H2O
Molecular Weight:
 137.86
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More