• USA Home
  • Search Results

  • sodiummethoxidesolution540212441411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium methoxide solution

3 Products
Synonym:   Sodium methylate
Image of Sodium methoxide solution
CAS Number:
Linear Formula:
 CH3ONa
Molecular Weight:
 54.02
 Beilstein/REAXYS Number:
 3592982
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Vapor pressure
  Special Grade
  Density
Show More