• USA Home
  • Search Results

  • sodiumnitrate15n85993143245811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium nitrate-15N

5 Products
Image of Sodium nitrate-<SUP>15</SUP>N
CAS Number:
Linear Formula:
 Na15NO3
Molecular Weight:
 85.99
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Molecular weight
Show More