• USA Home
  • Search Results

  • sodiumnitrate8499763199411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium nitrate

13 Products
Synonym:   Nitric acid sodium salt, Sodium nitrate
Image of Sodium nitrate
CAS Number:
Empirical Formula (Hill Notation):
 NNaO3
Molecular Weight:
 84.99
 EC Index Number
 231-554-3
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Quality clarifier
  Product Line
  Physical Form
  Titertest
  Potency
  Melting Point
  Density
  Bulk Density
  pH Value
  Solubility
  Assay
Show More