• USA Home
  • Search Results

  • sodiumnitrite6900763200011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium nitrite

11 Products
Image of Sodium nitrite
CAS Number:
Linear Formula:
 NaNO2
Molecular Weight:
 69.00
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Application(s)
  Quality clarifier
  Purity Grade
  Loss
  Potency
  Density
  Bulk Density
  Agency/Method
  Solubility
  Melting Point
  Product Line
  Vapor pressure
  Special Grade
  pH Value
  Physical Form
Show More