• USA Home
  • Search Results

  • sodiumnitroferricyanideiiidihydrate297951375538911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium nitroferricyanide(III) dihydrate

1 Products
Synonym:   Nitroprusside sodium, SNP, Sodium nitroprusside, Sodium pentacyanonitrosylferrate
Image of Sodium nitroferricyanide(III) dihydrate
CAS Number:
Linear Formula:
 Na2[Fe(CN)5NO] · 2H2O
Molecular Weight:
 297.95
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
  Physical Form
  Special Grade
Show More