• USA Home
  • Search Results

  • sodiumoxide6198131359311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium oxide

1 Products
Synonym:   Disodium monoxide, Disodium oxide
Image of Sodium oxide
CAS Number:
Linear Formula:
 Na2O
Molecular Weight:
 61.98
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
645613 Sodium oxide, 80%,