• USA Home
  • Search Results

  • sodiumparaperiodate293891394038011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium (para)periodate

1 Products
Image of Sodium (para)periodate
CAS Number:
Linear Formula:
 Na3H2IO6
Molecular Weight:
 293.89
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More