• USA Home
  • Search Results

  • sodiumpentafluoropropionate1860137877811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium pentafluoropropionate

1 Products
Synonym:   Pentafluoropropionic acid sodium salt, Perfluoropropionic acid sodium salt
Image of Sodium pentafluoropropionate
CAS Number:
Linear Formula:
 CF3CF2COONa
Molecular Weight:
 186.01
 Beilstein/REAXYS Number:
 3636648
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More