• USA Home
  • Search Results

  • sodiumperchloratemonohydrate14046779107311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium perchlorate monohydrate

3 Products
Image of Sodium perchlorate monohydrate
CAS Number:
Linear Formula:
 NaClO4 · H2O
Molecular Weight:
 140.46
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Purity Grade
  pH Value
  Special Grade
  UV-Absorption Values
Show More