• USA Home
  • Search Results

  • sodiumphosphatedibasic14196755879411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium phosphate dibasic

14 Products
Synonym:   sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate
Image of Sodium phosphate dibasic
CAS Number:
Linear Formula:
 Na2HPO4
Molecular Weight:
 141.96
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Suitability
  UV-Absorption Condition
  Restriction
  Quality clarifier
  Special Grade
  UV-Absorption Values
  Product Line
  Purity Grade
  Loss
  Physical Form
  Solubility
  pH Value
  Assay
Show More

Product #
Description
S3264 Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration),
S5136 Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%,
S9763 Sodium phosphate dibasic, ACS reagent, ≥99.0%,
71640 Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T),
04276 Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance),
255793 Sodium phosphate dibasic, 99.95% trace metals basis,
71636 Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T),
71642 Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T),
W239901 Sodium phosphate dibasic, FG,
795410 Sodium phosphate dibasic, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%,
RDD022 Sodium phosphate dibasic, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.0%,
S7907 Sodium phosphate dibasic, BioXtra, ≥99.0%,
S0876 Sodium phosphate dibasic, ReagentPlus®, ≥99.0%,
SX0723 Sodium Phosphate, Dibasic, anhydrous, HPLC, Meets ACS Specifications,