• USA Home
  • Search Results

  • sodiumphosphatedibasicdodecahydrate358141003932411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium phosphate dibasic dodecahydrate

4 Products
Synonym:   sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate
Image of Sodium phosphate dibasic dodecahydrate
CAS Number:
Linear Formula:
 Na2HPO4 · 12H2O
Molecular Weight:
 358.14
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  UV-Absorption Values
  Quality clarifier
  Restriction
  Agency/Method
  Physical Form
  Special Grade
  Solubility
  pH Value
Show More