• USA Home
  • Search Results

  • sodiumphosphatemonobasic11998755880711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium phosphate monobasic

9 Products
Synonym:   Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate
Image of Sodium phosphate monobasic
CAS Number:
Linear Formula:
 NaH2PO4
Molecular Weight:
 119.98
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  pK-Values
  Restriction
  Product Line
  Suitability
  Solubility
  Absorption
  UV-Absorption Values
  Physical Form
  pH Value
  Assay
Show More