• USA Home
  • Search Results

  • sodiumphosphatemonobasicdihydrate156011347235011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium phosphate monobasic dihydrate

3 Products
Synonym:   Sodium dihydrogen phosphate dihydrate
Image of Sodium phosphate monobasic dihydrate
CAS Number:
Linear Formula:
 NaH2PO4 · 2H2O
Molecular Weight:
 156.01
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Condition
  Quality clarifier
  Product Line
  Solubility
  UV-Absorption Values
  pH Value
  Special Grade
Show More