• USA Home
  • Search Results

  • sodiumphosphonoformatetribasichexahydrate300043415656411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium phosphonoformate tribasic hexahydrate

1 Products
Synonym:   Foscarnet, Phosphonoformic acid trisodium salt hexahydrate
Image of Sodium phosphonoformate tribasic hexahydrate
CAS Number:
Linear Formula:
 Na2O3PCO2Na · 6H2O
Molecular Weight:
 300.04
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More