• USA Home
  • Search Results

  • sodiumpropionate960613740611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium propionate

5 Products
Synonym:   Propionic acid sodium salt
Image of Sodium propionate
CAS Number:
Linear Formula:
 CH3CH2COONa
Molecular Weight:
 96.06
 Beilstein/REAXYS Number:
 3566934
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
  Special Grade
  Agency/Method
  Assay
Show More