• USA Home
  • Search Results

  • sodiumptoluenesulfinate1781882479311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium p-toluenesulfinate

2 Products
Synonym:   p-Toluenesulfinic acid sodium salt
Image of Sodium <I>p</I>-toluenesulfinate
CAS Number:
Linear Formula:
 4-CH3C6H4SO2Na
Molecular Weight:
 178.18
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More