• USA Home
  • Search Results

  • sodiumpyrophosphatetetrabasicdecahydrate446061347236111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium pyrophosphate tetrabasic decahydrate

2 Products
Synonym:   Pyrophosphate inorganic, Sodium diphosphate tetrabasic, Tetrasodium pyrophosphate
Image of Sodium pyrophosphate tetrabasic decahydrate
CAS Number:
Linear Formula:
 Na4P2O7 · 10H2O
Molecular Weight:
 446.06
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  pH Value
  UV-Absorption Values
  Solubility
  Product Line
Show More