• USA Home
  • Search Results

  • sodiumpyruvate1100411324611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium pyruvate

8 Products
Synonym:   α-Ketopropionic acid sodium salt, 2-Oxopropanoic acid sodium salt, Pyruvic acid sodium salt
Image of Sodium pyruvate
CAS Number:
Linear Formula:
 CH3COCOONa
Molecular Weight:
 110.04
 Beilstein/REAXYS Number:
 3568341
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Loss
  Biological Source
  Application(s)
  Product Line
  Special Grade
  Solubility
  Physical Form
Show More