• USA Home
  • Search Results

  • sodiumselenide12494131385511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium selenide

1 Products
Image of Sodium selenide
CAS Number:
Empirical Formula (Hill Notation):
 Na2Se
Molecular Weight:
 124.94
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
796948 Sodium selenide, 95%,