• USA Home
  • Search Results

  • sodiumselenite172941010218811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium selenite

4 Products
Image of Sodium selenite
CAS Number:
Linear Formula:
 Na2SeO3
Molecular Weight:
 172.94
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Special Grade
  Vapor pressure
  Product Line
  Application(s)
  Physical Form
Show More