• USA Home
  • Search Results

  • sodiumstearate3064682216211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium stearate

2 Products
Synonym:   Octadecanoic acid sodium salt, Stearic acid sodium salt
Image of Sodium stearate
CAS Number:
Linear Formula:
 CH3(CH2)16COONa
Molecular Weight:
 306.46
 Beilstein/REAXYS Number:
 3576813
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Ice Shipping
  Functional group
  Special Grade
  Physical Form
Show More