• USA Home
  • Search Results

  • sodiumsulfate14204775782611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium sulfate

24 Products
Image of Sodium sulfate
CAS Number:
Linear Formula:
 Na2SO4
Molecular Weight:
 142.04
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  pH Range
  Vapor pressure
  Ice Shipping
  Restriction
  Suitability
  Purity Grade
  Application(s)
  Quality clarifier
  UV-Absorption Values
  Potency
  Bulk Density
  Product Line
  Loss
  Solubility
  Agency/Method
  Physical Form
  Special Grade
  pH Value
Show More

Product #
Description
239313 Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular,
238597 Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder,
1.06649 Sodium sulfate, anhydrous for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
31481 Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%,
798592 Sodium sulfate, anhydrous, granular, free-flowing, Redi-Dri, ACS reagent, ≥99%,
13464 Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance),
746363 Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%,
71963 Sodium sulfate, tested according to Ph. Eur., anhydrous,
204447 Sodium sulfate, ≥99.99% trace metals basis,
S9627 Sodium sulfate, ReagentPlus®, ≥99.0%,
793531 Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%,
80948 Sodium sulfate, suitable for HPLC, LiChropur, 99.0-101.0% (T),
1.06639 Sodium sulfate, anhydrous granulated for organic trace analysis EMSURE®,
1.06637 Sodium sulfate, anhydrous, coarse granules for analysis EMSURE® ACS,
71959 Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T),
1.06647 Sodium sulfate, anhydrous 99.99 Suprapur®,
1.06645 Sodium sulfate, anhydrous fine powder EMPROVE® ESSENTIAL Ph Eur,BP,JPE,USP,
8.22286 Sodium sulfate, anhydrous for synthesis,
SX0760 Sodium sulfate, anhydrous, crystals, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
S5640 Sodium sulfate, ≥99.0%, suitable for plant cell culture,
1.06643 Sodium sulfate, anhydrous EMPROVE® ESSENTIAL Ph Eur,BP,USP,E 514,
S6547 Sodium sulfate, BioXtra, ≥99.0%,
1.37144 Sodium sulfate, anhydrous EMPROVE® EXPERT Ph Eur,USP,BP,
SX0760E Sodium sulfate, anhydrous, crystals, Meets ACS Specifications Tracepur®,