• USA Home
  • Search Results

  • sodiumsulfatedecahydrate32220772773311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium sulfate decahydrate

6 Products
Synonym:   Sodium sulfate decahydrate
Image of Sodium sulfate decahydrate
CAS Number:
Empirical Formula (Hill Notation):
 Na2O4S · 10H2O
Molecular Weight:
 322.20
 EC Index Number
 231-820-9
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Suitability
  UV-Absorption Values
  Solubility
  Quality clarifier
  Purity Grade
  Density
  Bulk Density
  Melting Point
  Loss
  pH Value
  Assay
Show More