• USA Home
  • Search Results

  • sodiumsulfidehydrate7804anhydrousbasis2761045311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium sulfide hydrate

1 Products
Image of Sodium sulfide hydrate
CAS Number:
Empirical Formula (Hill Notation):
 Na2S · xH2O
Molecular Weight:
 78.04 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More