• USA Home
  • Search Results

  • sodiumtartratedibasicdihydrate23008610624711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sodium tartrate dibasic dihydrate

2 Products
Synonym:   L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate
Image of Sodium tartrate dibasic dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C4H4Na2O6 · 2H2O
Molecular Weight:
 230.08
 Beilstein/REAXYS Number:
 6121732
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  UV-Absorption Values
  Solubility
  Product Line
Show More